siRNA Details
| Gene Symbol: Nkx2-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023504.1 | NP_075993.1 | 2 | 1049 | ||
| GenBank mRNA | AF202039.1 | - | 2 | 846 | ||
| AK005620.1 | - | 1 | 679 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkx2-4| Gene Symbol: Nkx2-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023504.1 | NP_075993.1 | 2 | 1049 | ||
| GenBank mRNA | AF202039.1 | - | 2 | 846 | ||
| AK005620.1 | - | 1 | 679 | |||