siRNA Details
| Gene Symbol: RNF41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242826.1 | NP_001229755.1 | 6 | 849 | ||
| NM_005785.3 | NP_005776.1 | 6 | 910 | |||
| NM_194358.2 | NP_919339.1 | 7 | 1053 | |||
| NM_194359.2 | NP_919340.1 | 6 | 805 | |||
| NR_040053.1 | 7 | 1053 | ||||
| XM_005268561.4 | XP_005268618.1 | 7 | 961 | |||
| XM_011537734.2 | XP_011536036.1 | 7 | 967 | |||
| XM_011537735.2 | XP_011536037.1 | 7 | 884 | |||
| GenBank mRNA | AF077599.1 | - | 5 | 743 | ||
| AK300752.1 | - | 7 | 911 | |||
| AK304576.1 | - | 7 | 750 | |||
| AK314811.1 | - | 6 | 769 | |||
| AY007109.1 | - | 1 | 28 | |||
| BC024284.1 | - | 7 | 1031 | |||
| BC032637.1 | - | 5 | 758 | 5496849 | ||
| BC036267.1 | - | 6 | 854 | |||
| BC095418.1 | - | 7 | 988 | |||
| BU170070.1 | - | 6 | 786 | |||

RNF41
Genomic Map