siRNA Details
| Gene Symbol: Ttc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028639.3 | NP_082915.1 | 20 | 3561 | ||
| GenBank mRNA | AK040578.1 | - | 20 | 3564 | ||
| AK040610.1 | - | 20 | 3532 | |||
| AK050263.1 | - | 20 | 3549 | |||
| AK080284.1 | - | 20 | 3540 | |||
| AK165244.1 | - | 13 | 2223 | |||
| BC023773.1 | - | 16 | 2519 | |||
| BC024549.1 | - | 1 | 185 | |||
| BC025613.1 | - | 2 | 1044 | |||
| BC042512.1 | - | 20 | 3238 | 4223801 | ||
| BC049254.1 | - | 4 | 1284 | |||
| BC057906.1 | - | 3 | 1184 | |||

Ttc7
Genomic Map