siRNA Details
| Gene Symbol: 4930402H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029432.1 | NP_083708.1 | 12, 13 | 1167 | ||
| GenBank mRNA | AK089462.1 | - | 12, 13 | 1191 | ||
| AK133294.1 | - | 12, 13 | 1309 | |||
| AK143611.1 | - | 12, 13 | 1192 | |||
| AK158367.1 | - | 12, 13 | 1189 | |||
| BC052447.1 | - | 12, 13 | 1167 | 6408647 | ||

4930402H24Rik
Genomic Map