siRNA Details
| Gene Symbol: 4930402H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029432.1 | NP_083708.1 | 37 | 4178 | ||
| GenBank mRNA | AK143611.1 | - | 37 | 4203 | ||
| BC030418.1 | - | 5 | 1096 | |||
| BC052447.1 | - | 37 | 4178 | 6408647 | ||

4930402H24Rik
Genomic Map