siRNA Details
| Gene Symbol: Clca3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139148.2 | NP_631887.2 | 2 | 302 | ||
| GenBank mRNA | AY008277.1 | - | 2 | 302 | ||
| BC058766.1 | - | 2 | 284 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clca3b| Gene Symbol: Clca3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139148.2 | NP_631887.2 | 2 | 302 | ||
| GenBank mRNA | AY008277.1 | - | 2 | 302 | ||
| BC058766.1 | - | 2 | 284 | |||