siRNA Details
| Gene Symbol: Rab40c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139154.2 | NP_631893.1 | 7 | 869 | ||
| GenBank mRNA | AB232639.1 | - | 6 | 763 | ||
| AF422144.1 | - | 6 | 822 | |||
| AK133766.1 | - | 6 | 645 | |||
| BC027008.1 | - | 7 | 857 | 5364768 | ||
| BC037461.1 | - | 8 | 1301 | |||

Rab40c
Genomic Map