siRNA Details
Gene Symbol: Catsper1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139301.2 | NP_647462.1 | 1, 2 | 926 | ||
GenBank mRNA | AF407332.1 | - | 1, 2 | 926 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Catsper1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139301.2 | NP_647462.1 | 1, 2 | 926 | ||
GenBank mRNA | AF407332.1 | - | 1, 2 | 926 |