siRNA Details
| Gene Symbol: Spice1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144550.4 | NP_653133.3 | 8 | 792 | ||
| GenBank mRNA | AK048789.1 | - | 8 | 736 | ||
| AK165031.1 | - | 6 | 635 | |||
| AY099108.1 | - | 8 | 1072 | |||
| BC052381.1 | - | 8 | 746 | 5703283 | ||
| BC056960.1 | - | 8 | 746 | 5703283 | ||
| CX568108.1 | - | 8 | 791 | |||

Spice1
Genomic Map