siRNA Details
| Gene Symbol: Depdc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144804.1 | NP_659053.1 | 9 | 1476 | ||
| GenBank mRNA | AK134830.1 | - | 9 | 1496 | ||
| AK144066.1 | - | 9 | 1489 | |||
| AK162086.1 | - | 9 | 1500 | |||
| AK162218.1 | - | 9 | 1499 | |||
| BC013499.1 | - | 9 | 1476 | 4222591 | ||

Depdc7
Genomic Map