siRNA Details
| Gene Symbol: PI4KA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058004.3 | NP_477352.3 | 39, 40 | 4777 | ||
| XM_005261634.1 | XP_005261691.1 | 38, 39 | 4684 | |||
| XM_017028829.1 | XP_016884318.1 | 38, 39 | 4711 | |||
| XM_017028830.1 | XP_016884319.1 | 25, 26 | 2933 | |||
| GenBank mRNA | AB210002.1 | - | 39, 40 | 4747 | ||
| AF012872.1 | - | 38, 39 | 4516 | |||
| AK302424.1 | - | 39, 40 | 4705 | |||
| AK314781.1 | - | 11, 12 | 1431 | |||
| BC018120.1 | - | 9, 10 | 1094 | |||
| BC053654.1 | - | 8, 9 | 1070 | |||
| L36151.1 | - | 7, 8 | 1063 | |||

PI4KA
Genomic Map