siRNA Details
| Gene Symbol: Heatr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144835.4 | NP_659084.3 | 5 | 717 | ||
| GenBank mRNA | AK044969.1 | - | 5 | 713 | ||
| AK087792.1 | - | 5 | 722 | |||
| AK143958.1 | - | 5 | 695 | |||
| AK163223.1 | - | 5 | 681 | |||
| AK165368.1 | - | 5 | 659 | |||
| AK165890.1 | - | 5 | 720 | |||
| AK172508.1 | - | 5 | 714 | |||
| BC024471.1 | - | 2 | 168 | |||

Heatr1
Genomic Map