siRNA Details
| Gene Symbol: Fam49b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144846.5 | NP_659095.1 | 6 | 639 | ||
| GenBank mRNA | AK028062.1 | - | 6 | 640 | ||
| AK075613.1 | - | 7 | 584 | |||
| AK183968.1 | - | 4 | 334 | |||
| AK205511.1 | - | 4 | 297 | |||
| BC011343.1 | - | 6 | 457 | 4012217 | ||

Fam49b
Genomic Map