siRNA Details
| Gene Symbol: Fam49b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144846.5 | NP_659095.1 | 12 | 2676 | ||
| GenBank mRNA | AK028062.1 | - | 12 | 2677 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam49b| Gene Symbol: Fam49b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144846.5 | NP_659095.1 | 12 | 2676 | ||
| GenBank mRNA | AK028062.1 | - | 12 | 2677 | ||