siRNA Details
| Gene Symbol: Ankrd54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144849.1 | NP_659098.1 | 2, 3 | 430 | ||
| GenBank mRNA | AK133785.1 | - | 2, 3 | 454 | ||
| AK143625.1 | - | 2, 3 | 457 | |||
| BC014743.1 | - | 2, 3 | 430 | 4503319 | ||
| DQ884878.1 | - | 2, 3 | 458 | |||

Ankrd54
Genomic Map