siRNA Details
| Gene Symbol: Slc7a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144852.3 | NP_659101.2 | 2 | 944 | ||
| GenBank mRNA | AK030586.1 | - | 2 | 948 | ||
| AK043772.1 | - | 2 | 945 | |||
| AK162398.1 | - | 2 | 988 | |||
| BC016100.1 | - | 2 | 1038 | 4911158 | ||

Slc7a4
Genomic Map