siRNA Details
| Gene Symbol: Cyyr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144853.3 | NP_659102.1 | 4 | 633 | ||
| GenBank mRNA | AF442733.1 | - | 4 | 368 | ||
| AK031840.1 | - | 4 | 657 | |||
| AY061854.1 | - | 4 | 655 | |||
| BC099957.1 | - | 4 | 633 | 6431603 | ||
| BC125594.1 | - | 4 | 427 | 40130109 | ||
| BC125596.1 | - | 4 | 427 | 40130111 | ||

Cyyr1
Genomic Map