siRNA Details
| Gene Symbol: Wdr36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110015.1 | NP_001103485.1 | 9 | 1117 | ||
| NM_001110016.1 | NP_001103486.1 | 9 | 1117 | |||
| NM_144863.4 | NP_659112.2 | 9 | 1117 | |||
| GenBank mRNA | AK041763.1 | - | 9 | 1037 | ||
| AK045396.1 | - | 9 | 1090 | |||
| AK077545.1 | - | 9 | 1046 | |||
| AK140019.1 | - | 9 | 1042 | |||
| AK155965.1 | - | 9 | 1016 | |||
| AK156822.1 | - | 1118 | ||||
| AK171683.1 | - | 9 | 1055 | |||
| AK172054.1 | - | 9 | 1042 | |||
| AK172377.1 | - | 9 | 1047 | |||
| AK214340.1 | - | 1 | 19 | |||
| BC022132.1 | - | 9 | 1089 | |||
| BC025809.1 | - | 9 | 1031 | |||

Wdr36
Genomic Map