siRNA Details
| Gene Symbol: Rpap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289569.1 | NP_001276498.1 | 12 | 1934 | ||
| NM_144911.3 | NP_659160.2 | 12 | 1920 | |||
| GenBank mRNA | AK034418.1 | - | 12 | 1936 | ||
| AK145830.1 | - | 12 | 1895 | |||
| BC021895.1 | - | 12 | 1881 | 3481916 | ||

Rpap2
Genomic Map