siRNA Details
| Gene Symbol: Tmem150a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144916.3 | NP_659165.1 | 8 | 1461 | ||
| GenBank mRNA | BC014685.1 | - | 7 | 1377 | 4241375 | |
| BC028825.1 | - | 8 | 1463 | 5097962 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem150a| Gene Symbol: Tmem150a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144916.3 | NP_659165.1 | 8 | 1461 | ||
| GenBank mRNA | BC014685.1 | - | 7 | 1377 | 4241375 | |
| BC028825.1 | - | 8 | 1463 | 5097962 | ||