siRNA Details
| Gene Symbol: Blvrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC027279.1 | - | 5 | 790 | 3492861 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Blvrb| Gene Symbol: Blvrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC027279.1 | - | 5 | 790 | 3492861 | |