siRNA Details
| Gene Symbol: Fam193b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145382.4 | NP_663357.2 | 8 | 3067 | ||
| GenBank mRNA | AK040568.1 | - | 2 | 1195 | ||
| AK155237.1 | - | 8 | 3061 | |||
| AK173303.1 | - | 8 | 1916 | |||
| BC021381.1 | - | 2 | 657 | 5043282 | ||
| BC023015.1 | - | 2 | 599 | 5364758 | ||
| BC025483.1 | - | 8 | 1926 | 5150988 | ||
| BC051255.1 | - | 2 | 3515 | |||

Fam193b
Genomic Map