siRNA Details
| Gene Symbol: BC016579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145389.2 | NP_663364.2 | 3, 4 | 483 | ||
| GenBank mRNA | AK078878.1 | - | 3, 4 | 434 | ||
| BC016579.1 | - | 3, 4 | 483 | 2615865 | ||
| CD552400.1 | - | 1, 2 | 122 | |||

BC016579
Genomic Map