siRNA Details
| Gene Symbol: Tapbpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145391.2 | NP_663366.2 | 7 | 1860 | ||
| GenBank mRNA | BC017613.1 | - | 7 | 1860 | 4912570 | |
| BC094656.1 | - | 7 | 1830 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tapbpl| Gene Symbol: Tapbpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145391.2 | NP_663366.2 | 7 | 1860 | ||
| GenBank mRNA | BC017613.1 | - | 7 | 1860 | 4912570 | |
| BC094656.1 | - | 7 | 1830 | |||