siRNA Details
| Gene Symbol: Chtf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145412.3 | NP_663387.3 | 4 | 2681 | ||
| GenBank mRNA | AK030924.1 | - | 4 | 2680 | ||
| AK088257.1 | - | 4 | 2690 | |||
| AK165993.1 | - | 2 | 2508 | |||
| AK193837.1 | - | 1 | 34 | |||
| BC023107.1 | - | 2 | 2453 | 3966700 | ||

Chtf8
Genomic Map