siRNA Details
| Gene Symbol: Rnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159624.1 | NP_001153096.1 | 8 | 1569 | ||
| NM_145417.3 | NP_663392.2 | 9 | 1686 | |||
| GenBank mRNA | AK031138.1 | - | 8 | 1509 | ||
| AK145466.1 | - | 9 | 1690 | |||
| AK159195.1 | - | 9 | 1629 | |||
| AK159445.1 | - | 9 | 1618 | |||
| AK159625.1 | - | 9 | 1629 | |||
| BC010520.1 | - | 9 | 1647 | |||
| BC019200.1 | - | 9 | 1657 | 5041005 | ||

Rnpep
Genomic Map