siRNA Details
| Gene Symbol: Rnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159624.1 | NP_001153096.1 | 10 | 1911 | ||
| NM_145417.3 | NP_663392.2 | 11 | 2028 | |||
| GenBank mRNA | AA271633.1 | - | 1 | 27 | ||
| AK031138.1 | - | 10 | 1851 | |||
| AK145466.1 | - | 11 | 2032 | |||
| AK159195.1 | - | 11 | 1971 | |||
| AK159445.1 | - | 11 | 1960 | |||
| AK159625.1 | - | 11 | 1971 | |||
| BC010520.1 | - | 11 | 1989 | |||
| BC019200.1 | - | 11 | 1999 | 5041005 | ||

Rnpep
Genomic Map