siRNA Details
| Gene Symbol: Ginm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145418.4 | NP_663393.1 | 8 | 1003 | ||
| GenBank mRNA | AK144481.1 | - | 8 | 967 | ||
| BC013529.1 | - | 8 | 1003 | 4238526 | ||
| BC024530.1 | - | 2 | 280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ginm1| Gene Symbol: Ginm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145418.4 | NP_663393.1 | 8 | 1003 | ||
| GenBank mRNA | AK144481.1 | - | 8 | 967 | ||
| BC013529.1 | - | 8 | 1003 | 4238526 | ||
| BC024530.1 | - | 2 | 280 | |||