siRNA Details
| Gene Symbol: Dhrs7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172112.1 | NP_001165583.1 | 2 | 259 | ||
| NM_145428.2 | NP_663403.1 | 2 | 230 | |||
| GenBank mRNA | AK052209.1 | - | 2 | 182 | ||
| AK139084.1 | - | 2 | 211 | |||
| AK144057.1 | - | 2 | 165 | |||
| AK205316.1 | - | 1 | 17 | |||
| BC003479.1 | - | 2 | 185 | 2655855 | ||
| JX196872.1 | - | 2 | 149 | |||

Dhrs7b
Genomic Map