siRNA Details
| Gene Symbol: Fam222b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145430.2 | NP_663405.1 | 3 | 2036 | ||
| GenBank mRNA | AK047860.1 | - | 3 | 1908 | ||
| AK085728.2 | - | 3 | 2057 | |||
| AK137590.1 | - | 1 | 1087 | |||
| AK140562.1 | - | 3 | 1909 | |||
| AK141119.1 | - | 3 | 1998 | |||
| BC017647.1 | - | 3 | 2140 | 4527119 | ||
| BC063099.1 | - | 2 | 1932 | 6393931 | ||

Fam222b
Genomic Map