siRNA Details
| Gene Symbol: Mfsd7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145447.2 | NP_663422.1 | 3 | 1265 | ||
| GenBank mRNA | AK145696.1 | - | 3 | 1222 | ||
| AY260574.1 | - | 3 | 1265 | |||
| AY260575.1 | - | 3 | 1265 | |||
| AY260576.1 | - | 3 | 1265 | |||
| BC011209.1 | - | 3 | 1265 | 4190027 | ||

Mfsd7c
Genomic Map