siRNA Details
| Gene Symbol: 9030617O03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145448.4 | NP_663423.2 | 4 | 544 | ||
| GenBank mRNA | AK033551.1 | - | 4 | 573 | ||
| AK038436.1 | - | 4 | 546 | |||
| AK081256.1 | - | 581 | ||||
| AK152622.1 | - | 4 | 573 | |||
| BC021385.1 | - | 5 | 529 | 5064508 | ||
| BC066161.1 | - | 5 | 618 | 30093441 | ||

9030617O03Rik
Genomic Map