siRNA Details
| Gene Symbol: Zfp503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145459.3 | NP_663434.2 | 3 | 3962 | ||
| GenBank mRNA | AK032903.1 | - | 2 | 2509 | ||
| AK046771.1 | - | 2 | 2979 | |||
| AY281290.1 | - | 2 | 3032 | |||
| AY281291.1 | - | 2 | 3006 | |||
| BC018484.1 | - | 3 | 1887 | 3481880 | ||
| BC058952.1 | - | 2 | 2879 | 5715786 | ||
| BC066027.1 | - | 3 | 3962 | 6401144 | ||

Zfp503
Genomic Map