siRNA Details
| Gene Symbol: Oxnad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145460.2 | NP_663435.2 | 8 | 1607 | ||
| GenBank mRNA | AK031978.1 | - | 7 | 1236 | ||
| AK032160.1 | - | 8 | 1410 | |||
| AK075924.1 | - | 7 | 1233 | |||
| AK139884.1 | - | 8 | 1607 | |||
| AK214280.1 | - | 1 | 249 | |||
| BC019806.1 | - | 8 | 1383 | 4018387 | ||

Oxnad1
Genomic Map