siRNA Details
| Gene Symbol: Shisa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145463.5 | NP_663438.1 | 2 | 2151 | ||
| GenBank mRNA | AB280737.1 | - | 2 | 1905 | ||
| AK034905.1 | - | 2 | 2126 | |||
| AK087512.1 | - | 2 | 2121 | |||
| BC024118.1 | - | 2 | 1589 | 5068886 | ||
| BC057640.1 | - | 2 | 1717 | 4018213 | ||

Shisa2
Genomic Map