siRNA Details
Gene Symbol: Ggact | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145466.2 | NP_663441.1 | 3 | 361 | ||
GenBank mRNA | AK050251.1 | - | 3 | 351 | ||
BC006662.1 | - | 3 | 331 | 3501534 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ggact | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145466.2 | NP_663441.1 | 3 | 361 | ||
GenBank mRNA | AK050251.1 | - | 3 | 351 | ||
BC006662.1 | - | 3 | 331 | 3501534 |