siRNA Details
| Gene Symbol: AKAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005858.3 | NP_005849.1 | 6 | 1051 | ||
| XM_011527624.2 | XP_011525926.1 | 6 | 1144 | |||
| XM_017026141.1 | XP_016881630.1 | 6 | 1196 | |||
| GenBank mRNA | AK303640.1 | - | 3 | 678 | ||
| AK308251.1 | - | 4 | 787 | |||
| AK309529.1 | - | 2 | 421 | |||
| BC037270.2 | - | 6 | 1041 | 4837309 | ||
| Y11997.1 | - | 6 | 1042 | |||

AKAP8
Genomic Map