siRNA Details
| Gene Symbol: Nipal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145469.5 | NP_663444.2 | 4, 5 | 442 | ||
| GenBank mRNA | AK033975.1 | - | 4, 5 | 443 | ||
| AK038556.1 | - | 4, 5 | 583 | |||
| AK079085.1 | - | 4, 5 | 875 | |||
| BC016107.1 | - | 4, 5 | 465 | 4921508 | ||
| BC030399.1 | - | 4, 5 | 959 | 4166825 | ||
| BC058207.1 | - | 4, 5 | 608 | 5361417 | ||

Nipal2
Genomic Map