siRNA Details
| Gene Symbol: Cerk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145475.4 | NP_663450.3 | 4 | 942 | ||
| GenBank mRNA | AB079067.1 | - | 4 | 449 | ||
| AK042077.1 | - | 4 | 533 | |||
| AK052269.1 | - | 4 | 708 | |||
| AK129416.1 | - | 1 | 84 | |||
| AK169528.1 | - | 4 | 527 | |||
| AK188338.1 | - | 2 | 172 | |||
| BC094253.1 | - | 4 | 941 | 6400872 | ||

Cerk
Genomic Map