siRNA Details
| Gene Symbol: Cerk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145475.4 | NP_663450.3 | 13 | 2556 | ||
| GenBank mRNA | AB079067.1 | - | 13 | 2064 | ||
| AK052269.1 | - | 13 | 2322 | |||
| AK129416.1 | - | 10 | 1699 | |||
| AK161801.1 | - | 1394 | ||||
| AK169528.1 | - | 13 | 2141 | |||
| BC094253.1 | - | 13 | 2555 | 6400872 | ||

Cerk
Genomic Map