siRNA Details
| Gene Symbol: Klhl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145479.4 | NP_663454.3 | 7 | 2489 | ||
| GenBank mRNA | AK028197.1 | - | 8 | 2634 | ||
| AK143514.1 | - | 7 | 2434 | |||
| AK190702.1 | - | 1 | 182 | |||
| AK191360.1 | - | 1 | 182 | |||
| AK204888.1 | - | 1 | 183 | |||
| AK211998.1 | - | 1 | 121 | |||
| BC005800.1 | - | 7 | 2467 | 2812589 | ||

Klhl22
Genomic Map