siRNA Details
| Gene Symbol: Fam160b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145505.4 | NP_663480.3 | 6 | 1096 | ||
| GenBank mRNA | AK029835.1 | - | 6 | 1426 | ||
| AK046518.1 | - | 6 | 1089 | |||
| AK054539.1 | - | 6 | 1044 | |||
| AK122530.1 | - | 4 | 575 | |||
| AK172373.1 | - | 6 | 1061 | |||
| BC062949.1 | - | 6 | 835 | 6839812 | ||

Fam160b1
Genomic Map