siRNA Details
| Gene Symbol: DSCR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005867.3 | NP_005858.1 | 3 | 472 | ||
| GenBank mRNA | AB000099.1 | - | 3 | 472 | ||
| BC069729.1 | - | 3 | 434 | 7262631 | ||
| BC096162.1 | - | 3 | 471 | 40003276 | ||
| BC096163.1 | - | 3 | 471 | 40003277 | ||
| BC096164.1 | - | 3 | 471 | 40003278 | ||
| DQ179113.1 | - | 3 | 422 | |||

DSCR4
Genomic Map