siRNA Details
| Gene Symbol: BC003331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077237.1 | NP_001070705.1 | 13 | 1891 | ||
| NM_145511.2 | NP_663486.2 | 14 | 1987 | |||
| GenBank mRNA | AK152022.1 | - | 11 | 1407 | ||
| AK152955.1 | - | 11 | 1408 | |||
| BC003331.1 | - | 12 | 1576 | 3256634 | ||

BC003331
Genomic Map