siRNA Details
| Gene Symbol: Wdr26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145514.5 | NP_663489.4 | 5, 6 | 1344 | ||
| GenBank mRNA | AK075743.1 | - | 5, 6 | 736 | ||
| AK082326.1 | - | 6, 7 | 639 | |||
| AK194474.1 | - | 1, 2 | 29 | |||
| BC020044.1 | - | 1, 2 | 89 | |||
| BC058601.1 | - | 3, 4 | 340 | |||
| BC094233.1 | - | 2, 3 | 229 | |||
| BC139336.1 | - | 5, 6 | 506 | 8862358 | ||
| BC139337.1 | - | 5, 6 | 506 | 8862359 | ||

Wdr26
Genomic Map