siRNA Details
| Gene Symbol: Farp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145519.2 | NP_663494.2 | 7 | 673 | ||
| GenBank mRNA | AK050860.1 | - | 7 | 633 | ||
| AK086699.1 | - | 7 | 667 | |||
| AK171713.1 | - | 7 | 699 | |||
| BC009153.1 | - | 7 | 662 | 2655209 | ||
| BI739824.1 | - | 5 | 298 | |||

Farp2
Genomic Map