siRNA Details
| Gene Symbol: Ppapdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145521.3 | NP_663496.1 | 2 | 1274 | ||
| GenBank mRNA | AK048016.1 | - | 3 | 902 | ||
| AK052883.1 | - | 2 | 1275 | |||
| AK134638.1 | - | 2 | 1398 | |||
| BC016136.1 | - | 2 | 1205 | 4910887 | ||

Ppapdc3
Genomic Map