siRNA Details
| Gene Symbol: Rabepk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145522.4 | NP_663497.2 | 4 | 500 | ||
| GenBank mRNA | AK035343.1 | - | 4 | 380 | ||
| AK153022.1 | - | 4 | 404 | |||
| AK206590.1 | - | 2 | 192 | |||
| BC019800.1 | - | 4 | 376 | 5150243 | ||

Rabepk
Genomic Map