siRNA Details
| Gene Symbol: Rabepk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145522.4 | NP_663497.2 | 8 | 1300 | ||
| GenBank mRNA | AK035343.1 | - | 8 | 1180 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rabepk| Gene Symbol: Rabepk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145522.4 | NP_663497.2 | 8 | 1300 | ||
| GenBank mRNA | AK035343.1 | - | 8 | 1180 | ||