siRNA Details
| Gene Symbol: Rabepk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145522.4 | NP_663497.2 | 5 | 677 | ||
| GenBank mRNA | AK018405.1 | - | 1 | 971 | ||
| AK035343.1 | - | 5 | 557 | |||
| AK153022.1 | - | 5 | 581 | |||
| BC019800.1 | - | 5 | 553 | 5150243 | ||

Rabepk
Genomic Map